Learn More
Medchemexpress LLC Benzenesulfonic acid, 4-amino-, sodium salt (1:1) | 515-74-2 | 100.0% | 195.17 g/mol | C6H6NNaO3S | 25 G

Supplier: Medchemexpress LLC 25GHYW042216
Sodium sulphanilate (sodium 4-aminobenzenesulfonate) is a water-soluble sulfonate salt used as a biochemical index of inflammation-associated acute-phase response and as a reagent in biochemical and analytical research. The solid reagent is supplied with COA and SDS documentation and is stored sealed at low temperature to limit moisture uptake.
- High purity (99.97%).
- Molecular weight 195.17 g/mol.
- Chemical formula C6H6NNaO3S.
- Supplied as a solid in a 25 G package.
- Storage: 4°C, sealed, away from moisture.
- Documentation: COA and SDS available.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.