Learn More
Medchemexpress LLC Benzenesulphonamide | 98-10-2 | MFCD00007930 | 99.4% | 157.19 g/mol | C6H7NO2S | 25 G
SDP
Supplier:  Medchemexpress LLC HY10108725G
Encompass_Preferred
Benzenesulphonamide is a small-molecule carbonic anhydrase inhibitor and analytical standard used in biochemical and analytical research. It is supplied as a white to off-white solid and as ready-to-use DMSO solutions for assay and reference purposes.
  • High assay purity (≈99.4%) for reliable analytical results.
  • Molecular formula C6H7NO2S; molecular weight 157.19 g/mol.
  • Available as a solid and 10 mM DMSO solutions for immediate use.
  • White to off-white solid suitable for laboratory handling and storage.
  • Used in assays targeting carbonic anhydrase activity and related research.
Catalog No. 50-002-13328