Learn More
Medchemexpress LLC Benzenesulphonamide | 98-10-2 | MFCD00007930 | 99.5% | 157.19 g/mol | C6H7NO2S | 1 ML

Supplier: Medchemexpress LLC HY10108710MM 1ML
Benzenesulphonamide is a carbonic anhydrase inhibitor supplied for research use. It is available as a ready-to-use 10 mM solution in DMSO (1 mL) and in solid form, and has reported CA II inhibitory activity and uses in studies of oxidative stress and amyloid-related pathways. For research use only.
- Carbonic anhydrase II inhibitory activity.
- Provided as a 10 mM solution in DMSO (1 mL) for ready-to-use assays.
- High purity (≈99.5% assay) suitable for research applications.
- Also available in solid quantities for synthesis or bulk use.
- Used in research on Alzheimer's disease, epilepsy, inflammation, and certain cancers.
- CAS number 98-10-2 for unambiguous identification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.