Learn More
Apexbio Technology LLC Benzethonium Chloride 121-54-0 50mg
SDP
Supplier:  Apexbio Technology LLC B167450
Encompass_Preferred
Benzethonium chloride (CAS 121-54-0) is a small-molecule inhibitor targeting neuronal nicotinic acetylcholine receptors (nAChRs) It is designed to modulate these ligand-gated ion channels thereby influencing neuronal signaling pathways Benzethonium chloride exerts its biological activity primarily through inhibition of nAChRs In experimental studies benzethonium chloride demonstrates inhibitory activity against both 4 2 and 7 subtypes of nAChRs with IC50 values of approximately 49 nM and 122 nM respectively Based on these pharmacological properties benzethonium chloride holds research potential in neuroscience and pharmacological studies exploring receptor-mediated neural transmission and nicotinic pathway regulation
Catalog No. 50-000-91270