Learn More
Selleck Chemical LLC Benzethonium Chloride S4162-1g
SDP
Supplier:  Selleck Chemical LLC S41621G
Encompass_Preferred
Benzethonium chloride is a potent inhibitor of nAChRs it inhibits 4 2 nAChRs and 7 nAChRs with IC50 of 49 nM and 122 nM respectively
Catalog No. 50-310-9125