Learn More
Medchemexpress LLC Benzethonium chloride (Standard) | 121-54-0 | 98.3% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0942R
Encompass_Preferred
Benzethonium chloride (Standard) serves as the analytical standard for Benzethonium (chloride) and is designated for research and analytical applications. This compound is known to inhibit nicotinic acetylcholine receptors in human recombinant α7 and α4β2 neurons within Xenopus laevis oocytes.
  • The compound is an analytical standard grade, utilized as a reference standard.
  • It is frequently applied in qualitative, quantitative, and methodological research experiments.
  • Its applications include techniques such as HPLC, GC, and MS.
Catalog No. 50-004-50480