Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzetimide hydrochloride | 5633-14-7 | 99.4% | 25 MG
SDP

Supplier:  Medchemexpress LLC 25MGHYB1547A

Encompass_Preferred

Benzetimide hydrochloride is a muscarinic acetylcholine receptor (mAChR) antagonist. This product is intended for research use only and is not for patient administration.

  • Target: mAChR
  • Purity: 99.44%
  • Molecular weight: 398.93
  • Formula: C23H27ClN2O2
  • Appearance: Solid
  • Color: White to off-white
  • Storage: 4°C, sealed storage, away from moisture. In solvent: -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture)
  • Solubility in DMSO: ≥ 32 mg/mL (80.21 mM)
  • Solubility in H2O: 16.67 mg/mL (41.79 mM)

Catalog No. 50-004-14171


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.