Learn More
Cayman Chemical BenzetImIdehydrochlorIde 50mg
SDP
Supplier:  Cayman Chemical 3452750 MG
Encompass_Preferred
A muscarinic acetylcholine receptor antagonist; induces mydriasis and inhibits pilocarpine-induced salivation and lacrimation in rats (ED50s = 0.06 and 0.04 mg/kg, respectively)
Catalog No. 50-295-0850