Learn More
Medchemexpress LLC Benzhydrol | 91-01-0 | 184.24 | 500 G
SDP
Supplier:  Medchemexpress LLC HYW004059500G
Encompass_Preferred
Benzhydrol (Diphenylmethanol) is a biochemical reagent used as a biological material or organic compound for life science-related research. It is for research use only.
  • Used as intermediates in pharmaceuticals (including antihistamines), agrochemicals, perfumes, and other organic compounds.
  • Used as a fixative in the perfume industry.
  • Involved in polymerization reactions as a terminating group.
  • Used as a precursor to prepare modafinil, benztropine, and diphehydramine.
Catalog No. 50-004-45872