Learn More
Medchemexpress LLC Benzisothiazolone | 2634-33-5 | MFCD00044001, MFCD00127753 | 99.8% | 151.19 g/mol | C7H5NOS | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW017401
Encompass_Preferred
Benzisothiazolone (1,2-benzisothiazol-3(2H)-one; CAS 2634-33-5) is a heterocyclic isothiazolone used as a biocide and preservative in industrial and laboratory contexts. It is supplied as a solid powder intended for formulation, analytical, and research use.
  • Broad-spectrum antimicrobial activity against bacteria and fungi.
  • Suitable as a preservative in aqueous and solvent-based systems.
  • Solid powder form for easy handling and formulation.
  • High purity for analytical and research applications (typical >99.8%).
  • Molecular formula C7H5NOS and molecular weight 151.19 g/mol.
Catalog No. 50-003-04105