Learn More
Selleck Chemical LLC Benznidazole S3741-25mg
SDP
Supplier:  Selleck Chemical LLC S374125MG
Encompass_Preferred
Benznidazole (Radanil) is a nitroimidazole derivative having an antiprotozoal activity by interfering with parasite protein biosynthesis influencing cytokines production and stimulating host phagocytosis
Catalog No. 50-310-9287