Learn More
Medchemexpress LLC Benzo[a]pyrene | 50-32-8 | MFCD00003602 | 252.31 g/mol | C20H12 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10737725MG
Encompass_Preferred
Benzo[a]pyrene is a polycyclic aromatic hydrocarbon used as a research chemical and analytical reference in toxicology and chemoprevention studies. It is a well-characterized carcinogen in animal models and is commonly used to investigate mechanisms of carcinogenesis, aryl hydrocarbon receptor (AhR) activation, and metabolic bioactivation.
  • Used as a reference standard for toxicology and analytical studies.
  • Applicable to chemoprevention and carcinogenesis research.
  • Molecular formula C20H12 and molecular weight 252.31 g/mol.
  • Solid form; store tightly sealed in a cool, well-ventilated area per supplier guidance.
  • Hazardous: mutagenic and carcinogenic; use appropriate protective measures and consult the SDS.
Catalog No. 50-002-41253