Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzo[a]pyrene | 50-32-8 | MFCD00003602 | 99.8% | 252.31 g/mol | C20H12 | 100 MG
SDP

Supplier:  Medchemexpress LLC HY107377100MG

Encompass_Preferred

Benzo[a]pyrene is an analytical reference standard (CAS 50-32-8) used for qualitative and quantitative analysis and for research into chemoprevention and carcinogenicity. Supplied as a solid with high purity, it is intended for laboratory research use only and not for clinical or human use.

  • High purity (99.76%).
  • Chemical formula C20H12.
  • Molecular weight 252.31 g/mol.
  • Suitable as a reference standard for HPLC, GC, and MS.
  • Commonly used in chemoprevention and carcinogenicity studies.
  • Shipped at room temperature; follow certificate of analysis for storage.

Catalog No. 50-002-41256


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.