Learn More
Cusabio Technology LLC Benzo a pyrene BAP ELISA Kit
Supplier:  Cusabio Technology LLC CSBE57B
Encompass_Preferred
This ELISA kit detects Benzo[a]pyrene (BAP) in vegetable oil samples including canola soybean peanut and corn oil with high sensitivity and specificity Designed for quantitative analysis it offers a detection range of 0-108 g/kg and a detection limit of 4 g/kg enabling precise measurement of BAP contamination at trace levels The kit employs a reliable immunoassay platform to deliver consistent results for food safety monitoring and quality control applications Its optimized protocol ensures minimal cross-reactivity with structurally similar compounds while maintaining efficient sample preparation workflows for oil matrices This ready-to-use solution provides laboratories with a standardized method to comply with regulatory requirements for polycyclic aromatic hydrocarbon (PAH) analysis in edible oils
Catalog No. 50-000-49913