Learn More
Medchemexpress LLC Benzo[b]thiophen-2-yl(5-bromo-2-fluorophenyl)methanol | 1034305-11-7 | 337.21 | 1 G
SDP
Supplier:  Medchemexpress LLC HYI05631G
Encompass_Preferred
Benzo[b]thiophen-2-yl(5-bromo-2-fluorophenyl)methanol is a chemical compound that serves as a drug intermediate for the synthesis of various active compounds.
  • Drug intermediate
  • Suitable for synthesizing active compounds
  • Store at 4°C, protected from light
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
  • For research use only
Catalog No. 50-004-29232