Learn More
Medchemexpress LLC Benzo[b]thiophene-2-carboxamide, N-[4-[4-(2-methoxyphenyl)-1-piperazinyl]butyl]- | 474432-65-0 | 99.9% | 423.57 g·mol⁻¹ | C24H29N3O2S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1388095MG
Encompass_Preferred
FAUC 346 is a small-molecule research compound characterized as a highly selective dopamine D3 receptor partial agonist. It shows potent D3 activity and is intended for in vitro pharmacology and receptor profiling studies, supplied as a high-purity analytical standard for research applications.
  • Highly selective dopamine D3 partial agonist with low-nanomolar potency (EC50 ≈ 1.5 nM, Ki ≈ 0.23 nM).
  • Some affinity for 5-HT1A receptors (Ki ≈ 41 nM), useful for selectivity studies.
  • Supplied as a high-purity research reagent (reported purity 99.9%).
  • Molecular formula C24H29N3O2S and molecular weight 423.57 g·mol⁻¹ for analytical reference.
  • Suitable for in vitro pharmacology, receptor binding, and screening workflows.
  • Small-quantity packaging appropriate for assay development and early-stage research.
Catalog No. 50-002-21002