Learn More
Medchemexpress LLC Benzo[b]thiophene-2-carboxamide, N-[4-[4-(2-methoxyphenyl)-1-piperazinyl]butyl]- | 474432-65-0 | 99.90% | 423.57 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY138809
Encompass_Preferred
FAUC 346 is a highly selective D3 partial agonist with an EC50 of 1.5 nM. It is intended for research use only.
  • Highly selective D3 partial agonist with an EC50 of 1.5 nM
  • D3-selective ligand with a Ki of 0.23 nM in CHO cells for the D3 receptor
  • Shows affinity for 5HT1A receptors (Ki = 41 nM) and α1 receptors (Ki = 15 nM)
Catalog No. 50-003-48418