Learn More
Medchemexpress LLC Benzo[b]thiophene-3-carbaldehyde | 5381-20-4 | 162.21 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0022511G
Encompass_Preferred
Benzo[b]thiophene-3-carbaldehyde is a 3-formyl-substituted benzo[b]thiophene derivative that functions as an oxidizable substrate. It converts to benzo[b]thiophene-3-carboxylic acid under liquid-phase conditions and exhibits lower oxidation difficulty compared to similar compounds. This compound can significantly increase reaction rates in the presence of initiators and is a key starting material for synthesizing compounds such as heteroaryl chalcones.
  • Acts as an oxidizable substrate
  • Converts to benzo[b]thiophene-3-carboxylic acid
  • Exhibits lower oxidation difficulty
  • Increases reaction rates with initiators
  • Key starting material for heteroaryl chalcones synthesis
Catalog No. 50-004-47634