Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzo[e]pyrene-d12 | 205440-82-0 | MFCD00143670 | 98.0% | 264.38 g/mol | C20D12 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY132860S5MG

Encompass_Preferred

Benzo[e]pyrene-d12 is the deuterium-labeled isotopologue of benzo[e]pyrene used as an internal standard and tracer in analytical chemistry. It provides a mass-shifted reference for accurate quantitation and method development by mass spectrometry.

  • Isotopically labeled (D12) for clear mass separation.
  • Molecular formula C20D12; molecular weight 264.38 g/mol.
  • CAS number 205440-82-0.
  • 98.0% chemical purity and 98 atom % deuterium enrichment.
  • Supplied in small milligram quantities for analytical use.
  • Suitable as an internal standard for quantitative mass spectrometry workflows.

Catalog No. 50-002-62268


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.