Learn More
Medchemexpress LLC Benzodioxolo[5,6-c]-1,3-dioxolo[4,5-i]phenanthridin-6-ol, 5b,6,7,12b,13,14-hexahydro-5b,13-dimethyl-, (5bR,6S,12bR)- | 18797-79-0 | 367.40 | 20 MG
SDP
Supplier:  Medchemexpress LLC HYN082620MG
Encompass_Preferred
Corynoline is an orally active acetylcholinesterase (AChE) inhibitor with an IC50 of 30.6 μM. It exhibits multiple activities including anti-inflammatory, antinociceptive, antitumor, and analgesic effects. It can be used in the research of diseases such as tumors and inflammatory pain.
  • Orally active acetylcholinesterase (AChE) inhibitor
  • Exhibits anti-inflammatory effects
  • Demonstrates antinociceptive and analgesic properties
  • Shows antitumor activity
  • Applicable for research into tumors and inflammatory pain
Catalog No. 50-004-17056