Learn More
Medchemexpress LLC Benzofuran-2-boronic acid | 98437-24-2 | MFCD00236019 | 161.95 | 25 G

Supplier: Medchemexpress LLC 25GHYW004116
Benzofuran-2-boronic acid, identified by CAS number 98437-24-2, is a chemical compound with the molecular formula C8H7BO3 and a molecular weight of 161.95. This product is a white to off-white solid, exhibiting a high purity of 99.76% as determined by High-Performance Liquid Chromatography (HPLC).
- Appearance: White to off-white solid
- Purity: 99.76% (HPLC)
- ¹H NMR spectrum consistent with structure
- For research use only
- Store at -20°C, sealed and away from moisture
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.