Learn More
Medchemexpress LLC Benzoic acid, 2,4-difluoro-d3 | 1219804-63-3 | 161.12 | 10 MG

Supplier: Medchemexpress LLC HYW002068S10MG
Benzoic acid, 2,4-difluoro-d3 is a stable isotope, supplied as a white to off-white solid. This compound is suitable for various laboratory applications requiring high purity and specific isotopic enrichment.
- High purity of 99% (HPLC)
- Isotopic enrichment of 98.6%
- Store powder at -20°C for 3 years
- Store in solvent at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.