Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzoic acid, 3-amino-4-hydroxy-, methyl ester | 536-25-4 | 167.16 | 25 G
SDP

Supplier:  Medchemexpress LLC 25GHYW005637

Encompass_Preferred

Orthocaine is a local agent that can suppress or relieve pain. It features:

  • Molecular formula: C8H9NO3
  • Appearance: Solid
  • Color: Light brown to brown
  • Storage: 4°C, protect from light. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light)
  • Solubility in DMSO: 100 mg/mL (598.23 mM), requires ultrasonic
  • Local anesthetic

Catalog No. 50-004-49327


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.