Learn More
Medchemexpress LLC Benzoic acid, 3-bromo- | 585-76-2 | MFCD00002487 | 99.9% | 201.02 | C7H5BrO2 | 250g
SDP
Supplier:  Medchemexpress LLC HYY0188250G
Encompass_Preferred
3-Bromobenzoic acid is an important intermediate that can be used in the synthesis of other active compounds 3-Bromobenzoic acid can react with a variety of electrophiles to generate a variety of 2-substituted-3-bromobenzoic acid derivatives 3-Bromobenzoic acid is also an effective corrosion inhibitor for aluminum in alkalized Persian Gulf seawater and NaOH solution[1][2]
Catalog No. 50-002-69842