Learn More
Medchemexpress LLC Benzoic acid, 3-formyl-2-hydroxy- | 610-04-8 | 98.5% | 166.13 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY41570
Encompass_Preferred
3-Formylsalicylic acid is a compound that functions as a noncompetitive estrone sulfatase inhibitor, with an IC50 value of 0.15 mM and a Ki value of 0.12 mM. It exhibits low acute toxicity.
  • Noncompetitive estrone sulfatase inhibitor
  • IC50 value of 0.15 mM
  • Ki value of 0.12 mM
  • Low acute toxicity
Catalog No. 50-004-02295