Learn More
Medchemexpress LLC Benzoic acid, 3-methoxy- | 586-38-9 | MFCD00002499 | 98.4% | 152.15 g/mol | C8H8O3 | 100g
SDP
Supplier:  Medchemexpress LLC HYY0760100G
Encompass_Preferred
3-Methoxybenzoic acid can form complexes with metals such as europium (III) and gadolinium (III) 3-Methoxybenzoic acid can be metabolized by anaerobic bacteria through O-demethylation mechanism[1][2]
Catalog No. 50-002-69830