Learn More
Medchemexpress LLC Benzoic acid, 4-[3-(dibutylamino)propoxy]-, hydrochloride (1:1) | 437651-44-0 | 99.3% | C18H30ClNO3 | 500 MG

Supplier: Medchemexpress LLC HY75572500MG
Dronedarone impurity 25 hydrochloride is identified as an impurity of Dronedarone (HY-75839). It is categorized as a Drug Intermediate, with its pathway listed as "Others."
- Impurity of dronedarone
- Categorized as a drug intermediate
- Molecular formula: C18H30ClNO3
- CAS number: 437651-44-0
- Purity: 99.33%
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.