Learn More
Medchemexpress LLC Benzoic acid, 4-(4,5-diphenyl-1H-imidazol-2-yl)- | 5496-35-5 | MFCD00999654 | 99.7% | 340.37 g/mol | C22H16N2O2 | 25 G
SDP
Supplier:  Medchemexpress LLC HY10307825G
Encompass_Preferred
I-XW-053 (4-(4,5-diphenyl-1H-imidazol-2-yl)benzoic acid, CAS 5496-35-5) is a small-molecule inhibitor of the HIV-1 capsid protein used in antiviral and capsid-targeting research. Supplied as a solid with supporting analytical documentation, it is intended for biochemical assays and compound characterization. Solid pack sizes include 250 mg-50 g, with 25 g commonly available.
  • High purity suitable for research (99.71%).
  • Molecular formula C22H16N2O2; molecular weight 340.37 g/mol.
  • Available in multiple solid pack sizes, including 25 g.
  • Supplied with COA and SDS for traceability.
  • Suitable for antiviral and capsid-targeting biochemical studies.
Catalog No. 50-002-46111