Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzoic acid, 4-[5-[(2-furanylmethyl)amino]-1,2,4-triazolo[4,3-c]pyrimidin-8-yl]- | 2467965-71-3 | MFCD32701933 | 99.9% | C17H13N5O3 | 10MG
SDP

Supplier:  Medchemexpress LLC HY13097910MG

Encompass_Preferred

EED226-COOH is an EED-targeting ligand derived from EED226 designed for PROTAC development. It binds the EED subunit of PRC2 and is intended for conjugation to E3 ligase ligands to generate targeted protein degraders. The compound is supplied as an off-white to light-yellow solid and is provided with analytical characterization for biochemical and cell-based research.

  • Purity 99.9% (analytical).
  • Molecular formula C17H13N5O3.
  • Molecular weight 335.32 g·mol-1.
  • Solubility in DMSO 125 mg/mL (requires ultrasonic treatment).
  • Storage powder: -20°C (long-term) or 4°C (shorter term).
  • Includes datasheet, 1H NMR, LC-MS, COA, and SDS documentation.

Catalog No. 50-002-59799


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.