Learn More
Medchemexpress LLC Benzoic acid, 4-amino-, hydrazide | 5351-17-7 | 99.96% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB088010MM/1ML
Encompass_Preferred
4-Aminobenzohydrazide is an irreversible myeloperoxidase (MPO) inhibitor with an IC50 of 0.3 μM. This compound induces an oxidative stress burst in neutrophils with an ROS IC50 of 43.6 μM and has been utilized in subacute stroke research.
  • Functions as an irreversible myeloperoxidase (MPO) inhibitor
  • Induces oxidative stress burst in neutrophils
  • Utilized in subacute stroke research
  • Reduces PMA-induced APF-dependent fluorescence signals in human neutrophils
  • Inhibits oxidative burst in PMA-stimulated human neutrophils
Catalog No. 50-004-08477