Learn More
Medchemexpress LLC Benzoic acid, 4-hydroxy-, 2-methylpropyl ester | 4247-02-3 | MFCD00020167 | 98.9% | 194.23 | C11H14O3 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01502610G
Encompass_Preferred
Isobutylparaben (isobutyl 4-hydroxybenzoate) is a research-grade analytical reagent used as a preservative and a biological probe. It is supplied as a solid for laboratory and analytical applications and is used in studies of receptor activation and cellular responses, including antimicrobial assays, apoptosis, and oxidative stress investigations.
  • High purity suitable for analytical use.
  • Solid form for easy weighing and storage.
  • Reported agonist activity for PXR, CAR, and PPAR.
  • Suitable for studies of bacterial activity, apoptosis, and reactive oxygen species.
  • Well-characterized molecular formula and molecular weight for analytical work.
Catalog No. 50-003-02608