Learn More
Medchemexpress LLC Benzoic acid, 4-hydroxy-, butyl ester | 94-26-8 | MFCD00016478 | 98.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB143110MM/1ML
Encompass_Preferred
Butylparaben is an organic compound that acts as a successful antimicrobial preservative in cosmetics and can also be utilized in active molecule suspensions and as a food flavoring additive. This product is intended for research use only.
  • Inhibits orcine fertilization, cleavage, and blastocyst formation rates.
  • Induces DNA damage, apoptosis, and autophagy in porcine oocytes.
  • Has no systemic toxicity in rats at tested doses.
Catalog No. 50-004-13523