Learn More
Medchemexpress LLC Benzoic acid, 5-[2-chloro-4-(trifluoromethyl)phenoxy]-2-nitro-, methyl ester | 50594-67-7 | 99.69% | 375.68 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY147365
Encompass_Preferred
Acifluorfen-methyl is an inhibitor of protoporphyrinogen oxidase (Protox), an enzyme involved in heme and chlorophyll biosynthesis. It functions as a photobleaching herbicide.
  • Inhibitor of protoporphyrinogen oxidase (Protox)
  • Inhibits heme and chlorophyll biosynthesis
  • Functions as a photobleaching herbicide
  • For research use only
Catalog No. 50-003-63266