Learn More
Medchemexpress LLC Benzoic acid, 5-(aminosulfonyl)-4-chloro-2-[(2-furanylmethyl)amino]- | 54-31-9 | 99.8% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB01355G
Encompass_Preferred
Furosemide is a potent and orally active inhibitor of Na+/K+/2Cl- (NKCC) cotransporter, NKCC1 and NKCC2. It also acts as a GABAA receptor antagonist, displaying 100-fold selectivity for α6-containing receptors over α1-containing receptors. It functions as a loop diuretic and is used in studies of congestive heart failure, hypertension, and edema.
  • Potent and orally active inhibitor of NKCC cotransporter
  • Antagonist of GABAA receptors with high selectivity for α6-containing receptors
  • Functions as a loop diuretic
  • Used for the study of congestive heart failure, hypertension, and edema
  • Targets NKCC and GABA receptor
  • Pathway involves membrane transporter/ion channel and neuronal signaling
Catalog No. 50-004-50508