Learn More
Medchemexpress LLC Benzoin methyl ether | 3524-62-7 | MFCD00008492 | 98.1% | 226.27 | C15H14O2 | 5 G

Supplier: Medchemexpress LLC HY1185715G
Benzoin methyl ether is a photoinitiator used in UV curing and photochemical research. It is supplied for laboratory use with a molecular formula of C15H14O2, molecular weight of 226.27 g/mol, and an assay around 98.1%. Follow recommended storage: solid at 4°C under nitrogen; solutions stored at -80°C for long-term or -20°C for short-term stability.
- Photoinitiator for UV curing and photochemical applications.
- High purity (~98.1%) suitable for research use.
- Available as small research pack for convenient handling.
- Molecular weight 226.27 g/mol; formula C15H14O2.
- Store under nitrogen at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.