Learn More
Sigma Aldrich Fine Chemicals Biosciences Benzonase Nuclease, 9025-65-4, MFCD00131010, 25 KU
Supplier:  Sigma Aldrich Fine Chemicals Biosciences E826325KU
Encompass
The enzyme has been used for an assay to detect the amount of Tf1 RNA that is protected from degradation by the nuclease, Benzonase. ≥250 units/μL, ≥99% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution, ultrapure grade.
Catalog No. NC0253403