Learn More
Medchemexpress LLC Benzonitrile, 2-fluoro-5-formyl- | 218301-22-5 | 99.7% | 149.12 | 10 G
SDP
Supplier:  Medchemexpress LLC HY2013810G
Encompass_Preferred
2-Fluoro-5-formylbenzonitrile is a chemical compound that serves as a drug intermediate for the synthesis of various active compounds. It appears as an off-white to light yellow solid.
  • Used in the synthesis of active compounds
  • Off-white to light yellow solid appearance
  • Store at 4°C under nitrogen
Catalog No. 50-004-01115