Learn More
Medchemexpress LLC Benzonitrile, 2-fluoro-5-formyl- | 218301-22-5 | 99.7% | 149.12 | 100 G

Supplier: Medchemexpress LLC HY20138100G
2-Fluoro-5-formylbenzonitrile is a drug intermediate used for the synthesis of various active compounds.
- Solid appearance
- Off-white to light yellow color
- High purity (99.67%)
- Suitable for the synthesis of active compounds
- For research use only
- Requires specific storage conditions (e.g., 4°C under nitrogen)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.