Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzophenone-2 (2,2',4,4'-tetrahydroxybenzophenone) | 131-55-5 | MFCD00002278 | 99.7% | 246.22 g/mol | C13H10O5 | 500 G
SDP

Supplier:  Medchemexpress LLC HYW036120500G

Encompass_Preferred

Benzophenone-2 (2,2',4,4'-tetrahydroxybenzophenone) is a benzophenone-class UV filter and research reagent used to study estrogenic and xenobiotic receptor activity. It is applied in in vitro assays, receptor interaction studies, and formulation research where a well-characterized UV-absorbing reference material is required.

  • CAS number 131-55-5.
  • Chemical formula C13H10O5.
  • Molecular weight 246.22 g/mol.
  • High purity (≈99.7%) suitable for research use.
  • Used in studies of estrogen receptor α and pregnane X receptor activity.
  • Solid form for laboratory handling and formulation testing.

Catalog No. 50-002-99355


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.