Learn More
Medchemexpress LLC (Benzothiazol-2-ylthio)methyl thiocyanate | 21564-17-0 | MFCD00072503 | 88.7% | 238.35 | C9H6N2S3 | 10 MG

Supplier: Medchemexpress LLC HY10142510MG
TCMTB (Benzothiazol-2-ylthio)methyl thiocyanate is an antifouling agent and an emerging contaminant supplied as a research chemical. Provided as a yellow to brown liquid, it is used in studies of environmental contamination, biocidal activity, and material preservation. Handle per safety data sheet and store under inert atmosphere.
- CAS number 21564-17-0.
- Molecular formula C9H6N2S3.
- Molecular weight 238.35 g/mol.
- Purity 88.7%.
- Physical state liquid; appearance yellow to brown.
- Recommended storage 4°C under nitrogen.
- Stability in solvent: -80°C (6 months), -20°C (1 month).
- For research use only; not for human or veterinary use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.