Learn More
Medchemexpress LLC Benzothiazole-5-boronic acid pinacol ester | 1073354-91-2 | MFCD09260439 | ≥98.0% | 261.15 | C13H16BNO2S | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0196331G
Encompass_Preferred
Benzothiazole-5-boronic acid pinacol ester is a boronate building block used in Suzuki-Miyaura cross-coupling reactions to introduce a benzothiazole moiety. It is provided as a research reagent with characterization data for laboratory synthesis and method development.
  • Suitable for Suzuki-Miyaura cross-coupling reactions.
  • Pinacol ester form offers enhanced stability and easy handling.
  • High purity (≥98.0%) supports reproducible results.
  • Molecular formula C13H16BNO2S and molecular weight 261.15 aid stoichiometry planning.
  • Available in multiple pack sizes for synthetic scale flexibility.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-003-01847