Learn More
Medchemexpress LLC Benzothiazole-5-boronic acid pinacol ester | 1073354-91-2 | MFCD09260439 | ≥98.0% | 261.15 | C13H16BNO2S | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0196335G
Encompass_Preferred
Benzothiazole-5-boronic acid pinacol ester is a boronate ester used as a building block in cross-coupling chemistry, primarily as the boronate partner in Suzuki-Miyaura reactions. It is supplied as a solid for research use only.
  • High purity (≥98.0%).
  • Molecular formula C13H16BNO2S and molecular weight 261.15.
  • White to off-white solid appearance.
  • Available in multiple package sizes, including 5 g.
  • Intended for research use only; not for clinical use.
Catalog No. 50-003-00270