Learn More
Medchemexpress LLC Benzothiazole-5-boronic acid pinacol ester | 1073354-91-2 | MFCD09260439 | ≥98.0% | 261.15 g/mol | C13H16BNO2S | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01963310G
Encompass_Preferred
Benzothiazole-5-boronic acid pinacol ester is an organoboron building block used as the boronate component in Suzuki-Miyaura cross-coupling reactions. Supplied for research use, it enables installation of benzothiazole groups in synthetic targets and is suitable for a range of organic synthesis workflows.
  • High purity (≥98.0%) for reproducible results.
  • Suitable boronate reagent for Suzuki-Miyaura cross-coupling.
  • Pinacol ester form increases chemical stability and handling convenience.
  • Molecular weight 261.15 g/mol and formula C13H16BNO2S for precise formulation.
  • Available in multiple pack sizes to support different reaction scales.
Catalog No. 50-003-01473