Learn More
Sigma Aldrich Fine Chemicals Biosciences Benzothiazole 96 FG25KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences W32560025KG
Encompass
Benzothiazole a volatile sulfur-containing heterocyclic compound is one of the key odorants of heated milk. It is also reported to occur as a flavor component of miso coconut meat and cooked-beef.
Catalog No. 50-290-7324