Learn More
Medchemexpress LLC Benzothioamide-d5 | 1219804-59-7 | 99.9% | 142.23 | C7H2D5NS | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW015801S5MG
Encompass_Preferred
Benzothioamide-d5 is the deuterium-labeled (d5) stable-isotope analog of benzothioamide supplied as a solid for use as a labeled reference, tracer, or internal standard in analytical and research applications.
  • Deuterium-labeled (d5) for mass-shift reference in isotopic studies.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS quantitative analyses.
  • Supplied as a solid for convenient storage and handling.
  • High reported chemical purity (99.9%) for reliable analytical performance.
  • Available in small precise quantities for method development and calibration.
Catalog No. 50-003-00525