Learn More
Medchemexpress LLC Benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate | 56602-33-6 | ≥99.0% | 442.28 g/mol | C12H22F6N6OP2 | 50g
SDP
Supplier:  Medchemexpress LLC HYY097350G
Encompass_Preferred
BOP (hexafluorophosphate) is a biochemical reagent that can be used as a biological material or organic compound for life science related research BOP hexafluorophosphate can be used in stepwise solid phase synthesis to prepare fragments of [Ala15]-GRF(1-29)-NH2[1]
Catalog No. 50-002-67737