Learn More
Cayman Chemical BenzoylhypaconIn 5mg
SDP
Supplier:  Cayman Chemical 340465 MG
Encompass_Preferred
A monoester diterpene alkaloid with anti-inflammatory activity; inhibits COX-2 activity in a cell-free assay (IC50 = 20.5 µM); reduces histamine-induced vascular permeability in rats at 200 mg/kg
Catalog No. 50-294-8149