Learn More
Enzo Life Sciences Benztropine . mesylate (1g). CAS: 132-17-2
SDP
Supplier:  Enzo Life Sciences ALX550143G001
Encompass_Preferred
Muscarinic acetylcholine receptor antagonist. Inhibits the dopamine transporter. Purity: ≥98% (HPLC). Solubility: Soluble in water. Long Term Storage: +4°C.
Catalog No. 50-200-9488