Learn More
Apexbio Technology LLC Benzydamine HCl 132-69-4 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B16775.1
Encompass_Preferred
Benzydamine hydrochloride (132-69-4) is a small-molecule inhibitor targeting cyclooxygenase (COX) enzymes It is designed to inhibit COX activity thereby reducing prostaglandin synthesis and regulating inflammatory pathways Benzydamine hydrochloride exerts its biological activity primarily through COX enzyme modulation and sodium channel blockade In cell-based assays benzydamine hydrochloride demonstrates prostaglandin production inhibition with an IC50 ranging from approximately 10 to 50 M depending on the experimental model Based on these pharmacological properties benzydamine hydrochloride holds research potential in investigating inflammatory pathways analgesic mechanisms and local anesthetic activity in vitro and in animal models
Catalog No. 50-000-94127