Learn More
Medchemexpress LLC Benzyl 2-acetamido-2-deoxy-α-D-galactopyranoside | 3554-93-6 | MFCD00057920 | 99.9% | 311.33 g/mol | C15H21NO6 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1293895MG
Encompass_Preferred
Benzyl-α-GalNAc is a small-molecule research reagent that functions as an O-glycosylation inhibitor. It is used in cell-based and in vivo studies to reduce mucin synthesis, modulate glycoprotein glycosylation, and investigate effects on fibrosis and tumor response to chemotherapy.
  • Inhibits O-glycosylation in cell and animal models.
  • Reduces mucin synthesis and alters glycoprotein processing.
  • Demonstrated anti-fibrotic effects by decreasing collagen expression in hepatic stellate cells.
  • Enhances chemotherapeutic response in preclinical tumor models.
  • High solubility in DMSO (≥100 mg/mL) and moderate solubility in water with warming (≈7 mg/mL).
  • High purity as supplied (COA reports 99.93% by HPLC).
  • Stable as powder under frozen storage conditions for long-term use (powder: -20°C for years).
Catalog No. 50-002-62188